Assay Details
| Gene Symbol: Hcfc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008224.4 | NP_032250.2 | 5 - 6 | 1147 | 63 | |
| GenBank mRNA | AJ627036.1 | - | 2 - 3 | 313 | 63 | |
| BC053742.1 | - | 5 - 6 | 1105 | 6509535 | 63 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AJ627036.1 transcripts.
Set Membership: |
|

Hcfc1
Genomic Map