Assay Details
| Gene Symbol: Hcls1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008225.2 | NP_032251.2 | 8 - 9 | 712 | 93 | |
| GenBank mRNA | AK088723.1 | - | 8 - 9 | 694 | 93 | |
| BC007469.1 | - | 8 - 9 | 682 | 3593456 | 93 | |
| D42120.1 | - | 8 - 9 | 643 | 93 | ||
| X84797.1 | - | 8 - 9 | 680 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hcls1
Genomic Map