Assay Details
| Gene Symbol: Hdgfrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK032572.1 | - | 6 - 7 | 1757 | 63 | |
| AK143616.1 | - | 8 - 9 | 1033 | 63 | ||
| AK144669.1 | - | 8 - 9 | 1040 | 63 | ||
| AK146813.1 | - | 8 - 9 | 1002 | 63 | ||
| AK146918.1 | - | 8 - 9 | 1010 | 63 | ||
| BC003741.1 | - | 8 - 9 | 999 | 3488803 | 63 | |
| D63850.1 | - | 8 - 9 | 1016 | 63 | ||
| FN687734.2 | - | 8 - 9 | 759 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hdgfrp2
Genomic Map