Assay Details
| Gene Symbol: Hdgfrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK032572.1 | - | 8 - 9 | 2153 | 52 | |
| AK143616.1 | - | 10 - 11 | 1429 | 52 | ||
| AK144669.1 | - | 10 - 11 | 1436 | 52 | ||
| AK146813.1 | - | 10 - 11 | 1398 | 52 | ||
| AK146918.1 | - | 10 - 11 | 1406 | 52 | ||
| BC003741.1 | - | 10 - 11 | 1395 | 3488803 | 52 | |
| D63850.1 | - | 10 - 11 | 1412 | 52 | ||
| FN687734.2 | - | 10 - 11 | 1155 | 52 | ||
More Information
Set Membership: |
|

Hdgfrp2
Genomic Map