Assay Details
| Gene Symbol: Hgs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159328.1 | NP_001152800.1 | 5 - 6 | 508 | 74 | |
| NM_008244.3 | NP_032270.3 | 5 - 6 | 508 | 74 | ||
| GenBank mRNA | AK046299.1 | - | 5 - 6 | 469 | 74 | |
| AK183070.1 | - | 1 - 2 | 59 | 74 | ||
| BC003239.1 | - | 5 - 6 | 452 | 3497966 | 74 | |
| D50050.1 | - | 5 - 6 | 458 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hgs
Genomic Map