Assay Details
Gene Symbol: Hiat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008246.2 | NP_032272.2 | 2 - 3 | 375 | 144 | |
GenBank mRNA | AK004779.1 | - | 2 - 3 | 314 | 144 | |
AK135865.1 | - | 2 - 3 | 196 | 144 | ||
AK158838.1 | - | 2 - 3 | 376 | 144 | ||
AK169626.1 | - | 2 - 3 | 315 | 144 | ||
BC118047.1 | - | 2 - 3 | 195 | 40106779 | 144 | |
D88315.1 | - | 2 - 3 | 248 | 144 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
