Assay Details
| Gene Symbol: Efs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010112.4 | NP_034242.2 | 2 - 3 | 1630 | 80 | |
| GenBank mRNA | AK030321.1 | - | 2 - 3 | 757 | 80 | |
| AK163191.1 | - | 2 - 3 | 671 | 80 | ||
| BC005438.1 | - | 2 - 3 | 699 | 3487600 | 80 | |
| BC031543.1 | - | 2 - 3 | 425 | 3669513 | 80 | |
| U28728.1 | - | 2 - 3 | 654 | 80 | ||
| U57686.1 | - | 2 - 3 | 354 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Efs
Genomic Map