Assay Details
| Gene Symbol: Ephx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010145.2 | NP_034275.1 | 2 - 3 | 261 | 69 | |
| GenBank mRNA | AK018249.1 | - | 2 - 3 | 270 | 69 | |
| BC029636.1 | - | 2 - 3 | 260 | 3491966 | 69 | |
| BC045194.1 | - | 2 - 3 | 261 | 4986422 | 69 | |
| BC057857.1 | - | 2 - 3 | 256 | 5067895 | 69 | |
| BC061493.1 | - | 2 - 3 | 287 | 4488885 | 69 | |
| U89491.1 | - | 2 - 3 | 230 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ephx1
Genomic Map