Assay Details
| Gene Symbol: H13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159551.1 | NP_001153023.1 | 9 - 10 | 972 | 70 | |
| NM_010376.4 | NP_034506.1 | 9 - 10 | 972 | 70 | ||
| GenBank mRNA | AF483214.1 | - | 9 - 10 | 939 | 70 | |
| AF515662.1 | - | 9 - 10 | 851 | 70 | ||
| AJ345032.1 | - | 9 - 10 | 982 | 70 | ||
| AK007664.1 | - | 9 - 10 | 954 | 70 | ||
| AK049101.1 | - | 9 - 10 | 946 | 70 | ||
| AK050664.1 | - | 9 - 10 | 915 | 70 | ||
| AK077983.1 | - | 2 - 3 | 67 | 70 | ||
| AK080966.1 | - | 9 - 10 | 934 | 70 | ||
| AK159176.1 | - | 9 - 10 | 946 | 70 | ||
| AK169922.1 | - | 9 - 10 | 929 | 70 | ||
| AK171790.1 | - | 8 - 9 | 775 | 70 | ||
| AK186920.1 | - | 3 - 4 | 171 | 70 | ||
| BC034217.1 | - | 4 - 5 | 246 | 70 | ||
| BC045195.1 | - | 5 - 6 | 328 | 70 | ||
| DQ168449.1 | - | 9 - 10 | 988 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

H13
Genomic Map