Assay Details
| Gene Symbol: Hap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010404.3 | NP_034534.1 | 3 - 4 | 648 | 90 | |
| NM_177981.2 | NP_817090.1 | 3 - 4 | 648 | 90 | ||
| GenBank mRNA | AJ000262.1 | - | 3 - 4 | 594 | 90 | |
| AJ002271.1 | - | 3 - 4 | 594 | 90 | ||
| AK138436.1 | - | 595 | 90 | |||
| BC034089.1 | - | 3 - 4 | 586 | 4481735 | 90 | |
| BC053043.1 | - | 3 - 4 | 635 | 5694628 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hap1
Genomic Map