Assay Details
| Gene Symbol: Ercc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011729.2 | NP_035859.2 | 9 - 10 | 2512 | 92 | |
| GenBank mRNA | AK137563.1 | - | 10 - 11 | 2344 | 92 | |
| AK140825.1 | - | 9 - 10 | 2341 | 92 | ||
| AK160581.1 | - | 3 - 4 | 1386 | 92 | ||
| AK160645.1 | - | 7 - 8 | 1884 | 92 | ||
| AK164205.1 | - | 9 - 10 | 2330 | 92 | ||
| BC033413.1 | - | 9 - 10 | 2232 | 92 | ||
| BC053034.1 | - | 9 - 10 | 2321 | 92 | ||
| BC092235.1 | - | 9 - 10 | 2303 | 92 | ||
| BC141070.1 | - | 9 - 10 | 2342 | 9055682 | 92 | |
| D16306.1 | - | 9 - 10 | 2207 | 92 | ||
| U40796.1 | - | 9 - 10 | 2408 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ercc5
Genomic Map