Assay Details
| Gene Symbol: Ets2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011809.3 | NP_035939.3 | 2 - 3 | 309 | 67 | |
| GenBank mRNA | AK014602.2 | - | 2 - 3 | 232 | 67 | |
| AK038348.1 | - | 2 - 3 | 296 | 67 | ||
| AK128933.1 | - | 2 - 3 | 261 | 67 | ||
| AK143687.1 | - | 2 - 3 | 261 | 67 | ||
| AK144974.1 | - | 2 - 3 | 260 | 67 | ||
| AK145852.1 | - | 2 - 3 | 145 | 67 | ||
| AK150175.1 | - | 2 - 3 | 239 | 67 | ||
| AK151315.1 | - | 2 - 3 | 260 | 67 | ||
| AK152750.1 | - | 2 - 3 | 262 | 67 | ||
| AK153339.1 | - | 2 - 3 | 261 | 67 | ||
| AK153478.1 | - | 2 - 3 | 261 | 67 | ||
| BC005486.1 | - | 2 - 3 | 230 | 3486021 | 67 | |
| BC005504.1 | - | 2 - 3 | 170 | 3490813 | 67 | |
| CT010235.1 | - | 1 - 2 | 71 | 67 | ||
| J04103.1 | - | 2 - 3 | 208 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ets2
Genomic Map