Assay Details
| Gene Symbol: Ern2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012016.2 | NP_036146.2 | 15 - 16 | 1830 | 60 | |
| GenBank mRNA | AF071777.1 | - | 15 - 16 | 1789 | 60 | |
| AK139291.1 | - | 15 - 16 | 1819 | 60 | ||
| AK153659.1 | - | 15 - 16 | 1820 | 60 | ||
| BC108999.2 | - | 15 - 16 | 1786 | 40048029 | 60 | |
| BC109000.1 | - | 15 - 16 | 1786 | 40048030 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ern2
Genomic Map