Assay Details
| Gene Symbol: Eno2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK131652.1 | - | 1 - 2 | 134 | ||
| BY282442.1 | - | 1 - 2 | 238 | 134 | ||
| CA751156.1 | - | 1 - 2 | 134 | |||
| X52380.1 | - | 1 - 2 | 134 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK131652.1, X52380.1 transcripts.
Set Membership: |
|

Eno2
Genomic Map