Assay Details
| Gene Symbol: Eno2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK003051.1 | - | 4 - 5 | 333 | 95 | |
| AK131652.1 | - | 4 - 5 | 372 | 95 | ||
| AK134571.1 | - | 3 - 4 | 256 | 95 | ||
| AK146655.1 | - | 4 - 5 | 338 | 95 | ||
| AK170169.1 | - | 3 - 4 | 253 | 95 | ||
| BC009018.1 | - | 4 - 5 | 308 | 95 | ||
| BC031739.1 | - | 4 - 5 | 326 | 4527274 | 95 | |
| CA751156.1 | - | 4 - 5 | 370 | 95 | ||
| X52380.1 | - | 4 - 5 | 351 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eno2
Genomic Map