Assay Details
| Gene Symbol: Eno2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK003051.1 | - | 11 - 12 | 1324 | 102 | |
| AK131652.1 | - | 11 - 12 | 1363 | 102 | ||
| AK134571.1 | - | 10 - 11 | 1246 | 102 | ||
| AK146655.1 | - | 11 - 12 | 1329 | 102 | ||
| AK170169.1 | - | 9 - 10 | 1487 | 102 | ||
| BC009018.1 | - | 9 - 10 | 942 | 102 | ||
| BC031739.1 | - | 11 - 12 | 1317 | 4527274 | 102 | |
| X52380.1 | - | 11 - 12 | 1342 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24816932 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24816931 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24816933 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019457 | BC043708.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019457 | X52380.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019457 | BC009018.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019457 | BC031739.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019457 | AK146655.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 79751 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544695 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 747616 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 891081 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 306080 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 145215 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170901 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019457 | BC043708.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019457 | X52380.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019457 | BC009018.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019457 | BC031739.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019457 | AK146655.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P748882 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P130028 | BC043708.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P130028 | X52380.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P130028 | BC009018.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P130028 | BC031739.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P130028 | AK146655.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24816925 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06056303 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06056292 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170901 | - | - |
| Whole Mouse Genome G4122A | A_51_P130028 | BC043708.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130028 | X52380.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130028 | BC009018.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130028 | BC031739.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130028 | AK146655.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P748882 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114691 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170901 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05688755 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24816933 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24816932 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24816931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24816852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05410237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05817355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816892 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05817352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05410241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05597481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05597491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05817360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816901 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24816910 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10547809 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547811 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547813 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547812 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547815 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547810 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547816 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547818 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547808 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547814 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547817 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418829_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10547807 | - | - |
| Mouse Genome 430 2.0 Array | 1418829_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1418829_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13807_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13807_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600024151.mm.1 | TC0600003116.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600024151.mm.1 | NM_013509.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600024154.mm.1 | TC0600003116.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600024154.mm.1 | NM_013509.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600024153.mm.1 | TC0600003116.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600024153.mm.1 | NM_013509.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600024152.mm.1 | TC0600003116.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600024152.mm.1 | NM_013509.3 | Refseq |
| Mouse Expression Set 430 | 1418829_PM_a_at | - | - |
| Mouse Expression Set 430 | 1418829_a_at | - | - |
| Murine Genome U74v2 Set | 99045_PM_at | - | - |
| Murine Genome U74v2 Set | 99045_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418829_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418829_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418829_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418829_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2320068 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2320068 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002320068 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2763002 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2763002 | - | - |
| mouse-6 v1.1 (Illumina) | scl013807.1_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2320068 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl013807.1_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2320068 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2763002 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2763002 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl013807.1_30-S | - | - |

Eno2
Genomic Map