Assay Details
| Gene Symbol: Epb4.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013513.2 | NP_038541.1 | 3 - 4 | 595 | 71 | |
| GenBank mRNA | AK133896.1 | - | 3 - 4 | 609 | 71 | |
| AK137484.1 | - | 3 - 4 | 676 | 71 | ||
| AK143841.1 | - | 3 - 4 | 575 | 71 | ||
| AK158219.1 | - | 3 - 4 | 513 | 71 | ||
| AK158247.1 | - | 3 - 4 | 559 | 71 | ||
| AK158370.1 | - | 3 - 4 | 522 | 71 | ||
| AK165288.1 | - | 3 - 4 | 522 | 71 | ||
| BC053739.1 | - | 3 - 4 | 566 | 71 | ||
| L35933.1 | - | 3 - 4 | 574 | 71 | ||
| U03487.1 | - | 3 - 4 | 525 | 71 | ||
| U04055.1 | - | 3 - 4 | 595 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epb4.2
Genomic Map