| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000014048.mm.1 |
NM_013904.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014048.mm.1 |
NM_013904.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014048.mm.1 |
TC1000001949.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000014044.mm.1 |
NM_013904.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014044.mm.1 |
NM_013904.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014044.mm.1 |
TC1000001949.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000014046.mm.1 |
NM_013904.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014046.mm.1 |
NM_013904.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014046.mm.1 |
TC1000001949.mm.1 |
MTA TC |