Assay Details
| Gene Symbol: Ebi3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015766.2 | NP_056581.1 | 1 - 2 | 308 | 56 | |
| GenBank mRNA | AF013114.1 | - | 1 - 2 | 291 | 56 | |
| AK155023.1 | - | 1 - 2 | 271 | 56 | ||
| AK155903.1 | - | 1 - 2 | 163 | 56 | ||
| BC008209.1 | - | 1 - 2 | 256 | 3154957 | 56 | |
| BY134465.1 | - | 1 - 2 | 309 | 56 | ||
| CX240802.1 | - | 1 - 2 | 255 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ebi3
Genomic Map