Assay Details
| Gene Symbol: Ebi3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015766.2 | NP_056581.1 | 4 - 5 | 771 | 108 | |
| GenBank mRNA | AF013114.1 | - | 4 - 5 | 754 | 108 | |
| AK155023.1 | - | 4 - 5 | 734 | 108 | ||
| AK155903.1 | - | 4 - 5 | 626 | 108 | ||
| AW495532.1 | - | 1 - 2 | 108 | |||
| BC008209.1 | - | 4 - 5 | 719 | 3154957 | 108 | |
| BI134394.1 | - | 1 - 2 | 108 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31252965 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31252967 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31252966 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202633 | AK155023.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202633 | AK155903.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202633 | AW495532.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202633 | BC008209.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202633 | BI134394.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202633 | AK155023.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202633 | AK155903.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202633 | AW495532.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202633 | BC008209.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202633 | BI134394.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 307951 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 56037 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84837 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202633 | AK155023.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202633 | AK155903.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202633 | AW495532.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202633 | BC008209.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202633 | BI134394.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202633 | AK155023.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202633 | AK155903.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202633 | AW495532.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202633 | BC008209.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202633 | BI134394.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31252946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31252950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31252953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31252961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31252947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31252970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31252963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31252964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31252966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31252959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31252956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31252957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31252948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31252967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31252969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31252958 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03338689 | - | - |
| Whole Mouse Genome G4122A | A_51_P202633 | AK155023.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P202633 | AK155903.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P202633 | AW495532.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P202633 | BC008209.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P202633 | BI134394.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P202633 | AK155023.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P202633 | AK155903.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P202633 | AW495532.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P202633 | BC008209.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P202633 | BI134394.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135118 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13531924 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13531921 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31252967 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31252965 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31252966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31252922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252965 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13092330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13176570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252960 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13553858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13540302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13576626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31252969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13540298 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10445978 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445979 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445982 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445981 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445980 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449222_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10445977 | - | - |
| Mouse Genome 430 2.0 Array | 1449222_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449222_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50498_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50498_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700009440.mm.1 | NM_015766.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009440.mm.1 | NM_015766.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009440.mm.1 | TC1700000992.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449222_at | - | - |
| Mouse Expression Set 430 | 1449222_PM_at | - | - |
| Murine Genome U74v2 Set | 93608_PM_at | - | - |
| Murine Genome U74v2 Set | 162357_at | - | - |
| Murine Genome U74v2 Set | 93608_at | - | - |
| Murine Genome U74v2 Set | 162357_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449222_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449222_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449222_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449222_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 664685 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5340300 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005340300 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2662322 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2662322 | - | - |
| mouse-6 v1.1 (Illumina) | scl50608.5.1_156-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5340300 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50608.5.1_156-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5340300 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2662322 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2662322 | - | - |

Ebi3
Genomic Map