Assay Details
| Gene Symbol: Elane | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015779.2 | NP_056594.2 | 3 - 4 | 384 | 70 | |
| GenBank mRNA | AK143710.1 | - | 3 - 4 | 70 | ||
| BC145800.1 | - | 3 - 4 | 449 | 40131103 | 70 | |
| DY245521.1 | - | 1 - 2 | 70 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
response to yeast
acute inflammatory response to antigenic stimulus
transcription from RNA polymerase II promoter
proteolysis
phagocytosis
response to UV
response to lipopolysaccharide
defense response to bacterium
negative regulation of growth of symbiont in host
negative regulation of chemokine biosynthetic process
negative regulation of interleukin-8 biosynthetic process
positive regulation of interleukin-8 biosynthetic process
positive regulation of smooth muscle cell proliferation
positive regulation of immune response
defense response to fungus
leukocyte migration
neutrophil mediated killing of fungus
RNA polymerase II transcription corepressor activity
protease binding
endopeptidase activity
serine-type endopeptidase activity
heparin binding
peptidase activity
serine-type peptidase activity
hydrolase activity
cytokine binding
negative regulation of transcription from RNA polymerase II promoter
response to yeast
acute inflammatory response to antigenic stimulus
transcription from RNA polymerase II promoter
proteolysis
phagocytosis
response to UV
response to lipopolysaccharide
defense response to bacterium
negative regulation of growth of symbiont in host
negative regulation of chemokine biosynthetic process
negative regulation of interleukin-8 biosynthetic process
positive regulation of interleukin-8 biosynthetic process
positive regulation of smooth muscle cell proliferation
positive regulation of immune response
defense response to fungus
leukocyte migration
neutrophil mediated killing of fungus
RNA polymerase II transcription corepressor activity
protease binding
endopeptidase activity
serine-type endopeptidase activity
heparin binding
peptidase activity
serine-type peptidase activity
hydrolase activity
cytokine binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P137219 | AK143710.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P137219 | NM_015779.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 41783 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819311 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119627 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120325 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P137219 | AK143710.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P137219 | NM_015779.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P144551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279490 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279491 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P120325 | - | - |
| Whole Mouse Genome G4122A | A_51_P144551 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P120325 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08865888 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08471652 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08865884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08865886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08563152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08841151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08843572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08843574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08843562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08865884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279485 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10364541 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364536 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364540 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364539 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364538 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364537 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422928_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10364535 | - | - |
| Mouse Genome 430 2.0 Array | 1422928_at | - | - |
| Mouse Genome 430 2.0 Array | 1422928_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50701_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50701_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000006027.mm.1 | NM_015779.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000006027.mm.1 | NM_015779.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000006027.mm.1 | TC1000000823.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422928_at | - | - |
| Mouse Expression Set 430 | 1422928_PM_at | - | - |
| Murine Genome U74v2 Set | 100755_PM_at | - | - |
| Murine Genome U74v2 Set | 100755_at | - | - |
| Murine Genome U74v2 Set | 171479_at | - | - |
| Murine Genome U74v2 Set | 171479_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422928_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422928_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422928_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422928_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 831777 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1570452 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001570452 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221700 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221700 | - | - |
| mouse-6 v1.1 (Illumina) | scl050701.4_101-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1570452 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl050701.4_101-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1570452 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1221700 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221700 | - | - |

Elane
Genomic Map