Assay Details
Gene Symbol: Hist1h1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015787.4 | NP_056602.1 | 1 | 56 | 51 | |
GenBank mRNA | BC089600.1 | - | 56 | 6745595 | 51 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC089600.1,NM_015787.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
