Assay Details
| Gene Symbol: Esd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285423.1 | NP_001272352.1 | 4 - 5 | 490 | 61 | |
| NM_016903.5 | NP_058599.1 | 4 - 5 | 357 | 61 | ||
| GenBank mRNA | AB025408.1 | - | 4 - 5 | 332 | 61 | |
| AK007527.1 | - | 4 - 5 | 345 | 61 | ||
| AK010683.1 | - | 4 - 5 | 362 | 61 | ||
| AK021209.1 | - | 4 - 5 | 285 | 61 | ||
| AK079131.1 | - | 4 - 5 | 310 | 61 | ||
| AK088115.1 | - | 4 - 5 | 405 | 61 | ||
| AK163074.1 | - | 4 - 5 | 285 | 61 | ||
| BC046766.1 | - | 4 - 5 | 282 | 5694094 | 61 | |
| BC096644.1 | - | 4 - 5 | 297 | 61 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Esd
Genomic Map