Assay Details
| Gene Symbol: Eif3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018749.2 | NP_061219.2 | 6 - 7 | 616 | 104 | |
| GenBank mRNA | AB012580.1 | - | 6 - 7 | 560 | 104 | |
| AK045228.1 | - | 6 - 7 | 558 | 104 | ||
| AK049767.1 | - | 6 - 7 | 555 | 104 | ||
| AK145689.1 | - | 6 - 7 | 571 | 104 | ||
| AK168256.1 | - | 6 - 7 | 606 | 104 | ||
| AK203325.1 | - | 236 | 104 | |||
| AK216723.1 | - | 2 - 3 | 149 | 104 | ||
| BC018598.1 | - | 6 - 7 | 554 | 104 | ||
| BC089020.1 | - | 6 - 7 | 550 | 30551991 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif3d
Genomic Map