Assay Details
| Gene Symbol: Eif3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018749.2 | NP_061219.2 | 12 - 13 | 1356 | 64 | |
| GenBank mRNA | AB012580.1 | - | 12 - 13 | 1297 | 64 | |
| AK045228.1 | - | 12 - 13 | 1298 | 64 | ||
| AK049767.1 | - | 12 - 13 | 1295 | 64 | ||
| AK145689.1 | - | 12 - 13 | 1311 | 64 | ||
| AK168256.1 | - | 12 - 13 | 1346 | 64 | ||
| BC018598.1 | - | 12 - 13 | 1294 | 64 | ||
| BC089020.1 | - | 12 - 13 | 1290 | 30551991 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif3d
Genomic Map