Assay Details
| Gene Symbol: Egfl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019397.3 | NP_062270.1 | 4 - 5 | 591 | 69 | |
| GenBank mRNA | AJ245672.1 | - | 4 - 5 | 593 | 69 | |
| AK053738.1 | - | 4 - 5 | 588 | 69 | ||
| AK078534.1 | - | 4 - 5 | 569 | 69 | ||
| BC117702.2 | - | 4 - 5 | 577 | 40062530 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Egfl6
Genomic Map