Assay Details
| Gene Symbol: Egfl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019397.3 | NP_062270.1 | 6 - 7 | 833 | 101 | |
| GenBank mRNA | AJ245672.1 | - | 6 - 7 | 835 | 101 | |
| AK053738.1 | - | 6 - 7 | 830 | 101 | ||
| AK078534.1 | - | 6 - 7 | 811 | 101 | ||
| BC117702.2 | - | 6 - 7 | 819 | 40062530 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Egfl6
Genomic Map