Assay Details
| Gene Symbol: Hao2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019545.4 | NP_062418.3 | 3 - 4 | 315 | 110 | |
| GenBank mRNA | AF272947.1 | - | 3 - 4 | 311 | 110 | |
| AJ251820.1 | - | 1 - 2 | 125 | 110 | ||
| AK018684.1 | - | 3 - 4 | 303 | 110 | ||
| AK078908.1 | - | 3 - 4 | 305 | 110 | ||
| AK143991.1 | - | 3 - 4 | 306 | 110 | ||
| BC027754.1 | - | 3 - 4 | 347 | 5064100 | 110 | |
More Information
Set Membership: |
|

Hao2
Genomic Map