Assay Details
| Gene Symbol: Hao2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019545.4 | NP_062418.3 | 4 - 5 | 474 | 80 | |
| GenBank mRNA | AF272947.1 | - | 4 - 5 | 470 | 80 | |
| AK018684.1 | - | 4 - 5 | 462 | 80 | ||
| AK078908.1 | - | 4 - 5 | 464 | 80 | ||
| AK143991.1 | - | 4 - 5 | 465 | 80 | ||
| BC027754.1 | - | 4 - 5 | 506 | 5064100 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hao2
Genomic Map