Assay Details
| Gene Symbol: Ecel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277925.1 | NP_001264854.1 | 4 - 5 | 1178 | 68 | |
| NM_021306.3 | NP_067281.2 | 4 - 5 | 1170 | 68 | ||
| GenBank mRNA | AB026294.1 | - | 3 - 4 | 998 | 68 | |
| AK039467.1 | - | 4 - 5 | 1170 | 68 | ||
| AK159365.1 | - | 4 - 5 | 1180 | 68 | ||
| BC057569.1 | - | 3 - 4 | 1000 | 6413731 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ecel1
Genomic Map