Assay Details
| Gene Symbol: Extl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163514.1 | NP_001156986.1 | 2 - 3 | 322 | 113 | |
| NM_021388.4 | NP_067363.3 | 2 - 3 | 515 | 113 | ||
| GenBank mRNA | AB032170.1 | - | 2 - 3 | 325 | 113 | |
| AB032171.1 | - | 2 - 3 | 310 | 113 | ||
| AF200973.1 | - | 2 - 3 | 287 | 113 | ||
| AK019370.1 | - | 2 - 3 | 253 | 113 | ||
| AK028677.1 | - | 2 - 3 | 488 | 113 | ||
| AK159281.1 | - | 2 - 3 | 502 | 113 | ||
| BC031438.1 | - | 2 - 3 | 479 | 4024555 | 113 | |
| BC094444.1 | - | 2 - 3 | 228 | 4507232 | 113 | |
| BG694725.1 | - | 2 - 3 | 515 | 113 | ||
| BY251302.1 | - | 2 - 3 | 322 | 113 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB032170.1,AB032171.1,AF200973.1,AK019370.1,AK028677.1,AK159281.1,BC031438.1,BC094444.1,NM_001163514.1,NM_021388.4 transcripts.
Set Membership: |
|

Extl2
Genomic Map