Assay Details
| Gene Symbol: Slc43a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021398.3 | NP_067373.2 | 7 - 8 | 1041 | 80 | |
| GenBank mRNA | AF188622.1 | - | 7 - 8 | 1035 | 80 | |
| BC089603.1 | - | 6 - 7 | 741 | 6758493 | 80 | |
| BU516072.1 | - | 1 - 2 | 77 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc43a3
Genomic Map