Assay Details
| Gene Symbol: Myoz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021503.2 | NP_067478.1 | 3 - 4 | 433 | 118 | |
| GenBank mRNA | AJ252148.1 | - | 3 - 4 | 395 | 118 | |
| AK003634.1 | - | 3 - 4 | 383 | 118 | ||
| AY013296.1 | - | 3 - 4 | 409 | 118 | ||
More Information
Set Membership: |
|

Myoz2
Genomic Map