Assay Details
| Gene Symbol: Erbb2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005868.2 | NP_001005868.1 | 25 - 26 | 4564 | 69 | |
| NM_001289473.1 | NP_001276402.1 | 24 - 25 | 4420 | 69 | ||
| NM_001289474.1 | NP_001276403.1 | 22 - 23 | 4096 | 69 | ||
| NM_001289475.1 | NP_001276404.1 | 24 - 25 | 4447 | 69 | ||
| GenBank mRNA | AF263743.1 | - | 4 - 5 | 1301 | 69 | |
| AK006453.1 | - | 2 - 3 | 144 | 69 | ||
| AK029054.1 | - | 14 - 15 | 3246 | 69 | ||
| AK051733.1 | - | 4 - 5 | 1079 | 69 | ||
| AK122473.1 | - | 24 - 25 | 4021 | 69 | ||
| BC005691.1 | - | 4 - 5 | 545 | 69 | ||
| BC028256.1 | - | 5 - 6 | 2007 | 69 | ||
| BC141178.1 | - | 22 - 23 | 3805 | 9055790 | 69 | |
| BC141188.1 | - | 25 - 26 | 4273 | 9055800 | 69 | |
| BC145498.1 | - | 24 - 25 | 4156 | 9054061 | 69 | |
| BC145509.1 | - | 24 - 25 | 4129 | 9054072 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Erbb2ip
Genomic Map