Assay Details
| Gene Symbol: Zfand6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022985.6 | NP_075361.2 | 4 - 5 | 486 | 117 | |
| GenBank mRNA | AJ251508.1 | - | 1190 | 117 | ||
| AK002775.1 | - | 4 - 5 | 487 | 117 | ||
| AK009403.1 | - | 3 - 4 | 344 | 117 | ||
| AK144902.1 | - | 3 - 4 | 336 | 117 | ||
| BC010683.1 | - | 3 - 4 | 347 | 2647905 | 117 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK014007.1 | - | Chr. - - [-] on Build | 118 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Zfand6
Genomic Map