Assay Details
| Gene Symbol: Mrps9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023514.4 | NP_076003.3 | 7 - 8 | 693 | 108 | |
| GenBank mRNA | AJ250193.1 | - | 3 - 4 | 108 | ||
| AK009082.1 | - | 7 - 8 | 643 | 108 | ||
| AK144656.1 | - | 7 - 8 | 651 | 108 | ||
| AK150985.1 | - | 7 - 8 | 650 | 108 | ||
| BC022587.1 | - | 2 - 3 | 131 | 108 | ||
| BC051936.1 | - | 6 - 7 | 386 | 108 | ||
| BC069980.1 | - | 5 - 6 | 287 | 108 | ||
| BC083159.1 | - | 6 - 7 | 454 | 108 | ||
| BC116437.2 | - | 7 - 8 | 643 | 40104493 | 108 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC022587.1 transcripts.
Set Membership: |
|

Mrps9
Genomic Map