Assay Details
| Gene Symbol: Nkiras1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023526.3 | NP_076015.2 | 2 - 3 | 296 | 91 | |
| GenBank mRNA | AF225707.1 | - | 2 - 3 | 153 | 91 | |
| AK028539.1 | - | 2 - 3 | 284 | 91 | ||
| AK148040.1 | - | 2 - 3 | 222 | 91 | ||
| AK167340.1 | - | 2 - 3 | 193 | 91 | ||
| BC061092.1 | - | 2 - 3 | 201 | 6589347 | 91 | |
| BY268943.1 | - | 2 - 3 | 297 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nkiras1
Genomic Map