Assay Details
| Gene Symbol: Slc44a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023557.3 | NP_076046.1 | 9 - 10 | 713 | 76 | |
| GenBank mRNA | AK145621.1 | - | 9 - 10 | 705 | 76 | |
| AK164684.1 | - | 8 - 9 | 1481 | 76 | ||
| AK166620.1 | - | 6 - 7 | 615 | 76 | ||
| BC010808.1 | - | 9 - 10 | 711 | 4240658 | 76 | |
| BC052652.1 | - | 9 - 10 | 711 | 30034507 | 76 | |
More Information
Set Membership: |
|

Slc44a4
Genomic Map