Assay Details
| Gene Symbol: Slc44a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023557.3 | NP_076046.1 | 15 - 16 | 1595 | 60 | |
| GenBank mRNA | AK008864.1 | - | 4 - 5 | 557 | 60 | |
| AK145621.1 | - | 15 - 16 | 1587 | 60 | ||
| AK164684.1 | - | 14 - 15 | 2363 | 60 | ||
| AK166620.1 | - | 12 - 13 | 1497 | 60 | ||
| BC010808.1 | - | 15 - 16 | 1593 | 4240658 | 60 | |
| BC052652.1 | - | 15 - 16 | 1593 | 30034507 | 60 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc44a4
Genomic Map