Assay Details
| Gene Symbol: Wdr31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290521.1 | NP_001277450.1 | 7 - 8 | 879 | 79 | |
| NM_001290522.1 | NP_001277451.1 | 7 - 8 | 769 | 79 | ||
| NM_023597.2 | NP_076086.1 | 7 - 8 | 772 | 79 | ||
| GenBank mRNA | AF274321.1 | - | 7 - 8 | 749 | 79 | |
| AK017259.1 | - | 7 - 8 | 762 | 79 | ||
| AK141026.1 | - | 7 - 8 | 877 | 79 | ||
| BC026500.1 | - | 6 - 7 | 658 | 79 | ||
| BC037102.1 | - | 7 - 8 | 762 | 5364307 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr31
Genomic Map