Assay Details
| Gene Symbol: Wdr31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290521.1 | NP_001277450.1 | 9 - 10 | 1087 | 89 | |
| NM_001290522.1 | NP_001277451.1 | 9 - 10 | 977 | 89 | ||
| NM_023597.2 | NP_076086.1 | 9 - 10 | 980 | 89 | ||
| GenBank mRNA | AF274321.1 | - | 9 - 10 | 957 | 89 | |
| AK017259.1 | - | 9 - 10 | 970 | 89 | ||
| AK141026.1 | - | 9 - 10 | 1085 | 89 | ||
| BC026500.1 | - | 8 - 9 | 866 | 89 | ||
| BC037102.1 | - | 9 - 10 | 970 | 5364307 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr31
Genomic Map