Assay Details
| Gene Symbol: Rgl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023622.4 | NP_076111.2 | 8 - 9 | 1158 | 120 | |
| GenBank mRNA | AF237669.1 | - | 8 - 9 | 1117 | 120 | |
| AF239661.1 | - | 8 - 9 | 1159 | 120 | ||
| AK004876.1 | - | 7 - 8 | 891 | 120 | ||
| AK053550.1 | - | 8 - 9 | 1145 | 120 | ||
| AK131140.1 | - | 664 | 120 | |||
| AK134654.1 | - | 1140 | 120 | |||
| AK165023.1 | - | 8 - 9 | 1146 | 120 | ||
| AK165746.1 | - | 8 - 9 | 1152 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rgl3
Genomic Map