Assay Details
| Gene Symbol: Rgl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023622.4 | NP_076111.2 | 11 - 12 | 1378 | 83 | |
| GenBank mRNA | AF237669.1 | - | 11 - 12 | 1337 | 83 | |
| AF239661.1 | - | 11 - 12 | 1379 | 83 | ||
| AK053550.1 | - | 11 - 12 | 1365 | 83 | ||
| AK131140.1 | - | 884 | 83 | |||
| AK134654.1 | - | 1360 | 83 | |||
More Information
Set Membership: |
|

Rgl3
Genomic Map