Assay Details
| Gene Symbol: Isyna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023627.1 | NP_076116.1 | 4 - 5 | 488 | 70 | |
| GenBank mRNA | AF288525.1 | - | 4 - 5 | 488 | 70 | |
| AK005029.2 | - | 4 - 5 | 466 | 70 | ||
| AK079323.1 | - | 4 - 5 | 465 | 70 | ||
| AK195643.1 | - | 1 - 2 | 68 | 70 | ||
| BC003458.1 | - | 4 - 5 | 479 | 2648362 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Isyna1
Genomic Map