Assay Details
| Gene Symbol: Spock3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252620.1 | NP_001239549.1 | 8 - 9 | 783 | 95 | |
| NM_001252621.1 | NP_001239550.1 | 7 - 8 | 774 | 95 | ||
| NM_023689.3 | NP_076178.1 | 8 - 9 | 897 | 95 | ||
| GenBank mRNA | AJ278998.1 | - | 8 - 9 | 791 | 95 | |
| AK013644.1 | - | 8 - 9 | 826 | 95 | ||
| AK049843.1 | - | 7 - 8 | 775 | 95 | ||
| AK140736.1 | - | 1 - 2 | 84 | 95 | ||
| AK220280.1 | - | 8 - 9 | 881 | 95 | ||
| BC017601.1 | - | 8 - 9 | 796 | 4502169 | 95 | |
| BC053334.1 | - | 8 - 9 | 768 | 6492113 | 95 | |
More Information
Set Membership: |
|

Spock3
Genomic Map