Assay Details
| Gene Symbol: Abcb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023732.2 | NP_076221.1 | 16 - 17 | 2418 | 100 | |
| GenBank mRNA | AK004605.1 | - | 16 - 17 | 2498 | 100 | |
| AK151165.1 | - | 16 - 17 | 2527 | 100 | ||
| AK164003.1 | - | 13 - 14 | 3336 | 100 | ||
| AK168642.1 | - | 16 - 17 | 2519 | 100 | ||
| BC006634.1 | - | 16 - 17 | 2418 | 3710552 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcb6
Genomic Map