Assay Details
| Gene Symbol: 1700020D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023781.5 | NP_076270.3 | 1 | 490 | 71 | |
| GenBank mRNA | AF319980.1 | - | 1 | 454 | 71 | |
| AK006150.1 | - | 1 | 491 | 71 | ||
| BC049585.1 | - | 1 | 487 | 6772543 | 71 | |
| BC061091.1 | - | 1 | 488 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31929125 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31929124 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31929123 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407866 | AK006150.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407866 | BC049585.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407866 | AF319980.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407866 | BC061091.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407866 | NM_023781.5 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407866 | AK006150.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407866 | BC049585.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407866 | AF319980.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407866 | BC061091.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407866 | NM_023781.5 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407866 | AK006150.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407866 | BC049585.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407866 | AF319980.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407866 | BC061091.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407866 | NM_023781.5 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407866 | AK006150.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407866 | BC049585.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407866 | AF319980.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407866 | BC061091.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407866 | NM_023781.5 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P407866 | AK006150.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P407866 | BC049585.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P407866 | AF319980.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P407866 | BC061091.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P407866 | NM_023781.5 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P407866 | AK006150.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P407866 | BC049585.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P407866 | AF319980.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P407866 | BC061091.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P407866 | NM_023781.5 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929155 | - | - |
| Whole Mouse Genome G4122A | A_51_P407866 | AK006150.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407866 | BC049585.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407866 | AF319980.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407866 | BC061091.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407866 | NM_023781.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407866 | AK006150.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407866 | BC049585.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407866 | AF319980.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407866 | BC061091.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407866 | NM_023781.5 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122029 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14492675 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14469774 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14469781 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14290702 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14469779 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31929124 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31929125 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31929123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14492697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14450630 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14140786 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10465104 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465105 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422633_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10465103 | - | - |
| Mouse Genome 430 2.0 Array | 1422633_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422633_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75555_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75555_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900008677.mm.1 | NM_023781.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900008677.mm.1 | NM_023781.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900008677.mm.1 | TC1900000972.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422633_PM_at | - | - |
| Mouse Expression Set 430 | 1422633_at | - | - |
| Murine Genome U74v2 Set | 165564_at | - | - |
| Murine Genome U74v2 Set | 165564_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422633_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422633_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422633_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422633_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 351073 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2970239 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002970239 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2606556 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2606556 | - | - |
| mouse-6 v1.1 (Illumina) | scl53499.1.4_164-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2970239 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53499.1.4_164-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2970239 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2606555 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2606555 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl53499.1.4_164-S | - | - |

1700020D05Rik
Genomic Map