Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200024578.mm.1 |
NM_024185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024578.mm.1 |
NM_024185.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024578.mm.1 |
TC0200002974.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024573.mm.1 |
NM_024185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024573.mm.1 |
NM_024185.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024573.mm.1 |
TC0200002974.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024577.mm.1 |
NM_024185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024577.mm.1 |
NM_024185.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024577.mm.1 |
TC0200002974.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024579.mm.1 |
NM_024185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024579.mm.1 |
NM_024185.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024579.mm.1 |
TC0200002974.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024580.mm.1 |
NM_024185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024580.mm.1 |
NM_024185.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024580.mm.1 |
TC0200002974.mm.1 |
MTA TC |