Assay Details
| Gene Symbol: Fam188a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024185.4 | NP_077147.2 | 4 - 5 | 615 | 65 | |
| GenBank mRNA | AK002900.1 | - | 4 - 5 | 566 | 65 | |
| AK009891.1 | - | 4 - 5 | 594 | 65 | ||
| AK010050.1 | - | 4 - 5 | 591 | 65 | ||
| AK049694.1 | - | 4 - 5 | 597 | 65 | ||
| AK143250.1 | - | 4 - 5 | 587 | 65 | ||
| BC002225.1 | - | 4 - 5 | 597 | 65 | ||
| BC027202.1 | - | 4 - 5 | 519 | 4217391 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam188a
Genomic Map