Assay Details
| Gene Symbol: Pcyt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024229.2 | NP_077191.2 | 8 - 9 | 768 | 66 | |
| GenBank mRNA | AK140458.1 | - | 8 - 9 | 808 | 66 | |
| AK144544.1 | - | 8 - 9 | 817 | 66 | ||
| AK166319.1 | - | 7 - 8 | 743 | 66 | ||
| BC003473.1 | - | 8 - 9 | 774 | 2650077 | 66 | |
| BC008276.1 | - | 8 - 9 | 768 | 3707732 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcyt2
Genomic Map